Learn More
Type IIR 3-ply Disposable Face Mask
Click to view available options
:
Type IIR Disposable Face Mask; Style: Type IIR
Description
- Disposable, single-use protection
- A CE Class I Medical Device, certified under (EU) 2017/745, meeting the EN14683:2019 standard for medical face masks
- Features a soft, flexible, internally wired upper seam for a comfortable snug fit around the nose and knitted elastic ear loops
- Nonwoven polypropylene inner and outer layers
- Melt blown polypropylene middle layer
- Meets the EN ISO 10993-10:2013 standard for skin irritation and sensitization
- Blue exterior
- Universal size and easy fitting
Specifications
Specifications
| Product Type | General Purpose Face Mask |
| Color | Blue |
| Material | Polypropylene |
| Size | 17.5 x 9.5 cm |
| Sterility | Non-sterile |
| Style | Pleated |
| Certifications/Compliance (Europe) | Medical Device Regulation: (EU) 2017/745; EN 14683:2019 |
| Disposable | Single-use |
| For Use With (Application) | Medical |
| Latex Free | Yes |
| Show More |