Learn More
GUCD1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82884-0.1ML
This item is not returnable.
View return policy
Description
GUCD1 Polyclonal specifically detects GUCD1 in Human samples. It is validated for Western Blot.
Specifications
| GUCD1 | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 83606 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| chromosome 22 open reading frame 13, LLN4 | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of Human C22orf13 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |