Learn More
CBLB Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-49920
This item is not returnable.
View return policy
Description
CBLB Polyclonal specifically detects CBLB in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| CBLB | |
| Immunoprecipitation | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate | |
| CBLB |