Learn More
Human VSIG8 Alexa Fluor 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems FAB9418S-100UG
Product Code. 18788648
  • 488.00 € / 100µg
Description
VSIG8 Monoclonal specifically detects VSIG8 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VSIG8
Monoclonal
Alexa Fluor 750
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
V-set and immunoglobulin domain containing 8
Human embryonic kidney cell line HEK293-derived transfected with human VSIG8 Met1-Gly263 Accession # Q5VU13
100 μg
391123
Human
Supernatant
Flow Cytometry
961823
Flow Cytometry 0.25-1 ug/10^6 cells
Q5VU13
Mouse
Protein A or G purified from hybridoma culture supernatant
Primary
Detects human VSIG8 in direct ELISAs. Stains human VSIG8 transfectants but not irrelevant transfectants in flow cytometry.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.