Learn More
Vimentin Antibody (CL0157) [Allophycocyanin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52867APC
Product Code. 30290284
  • 748.00 € / 0.10mL
Description
Vimentin Monoclonal antibody specifically detects Vimentin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Gene Knock-Out
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Vimentin
Monoclonal
APC
epididymis secretory sperm binding protein, FLJ36605, VIM, vimentin
This antibody was developed using a recombinant protein derived from P08670, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Cell Biology, Cellular Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Stem Cells
7431
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Gene Knock-Out
CL0157
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified