missing translation for 'onlineSavingsMsg'
Learn More

Vinylmagnesium bromide, 0.7M solution in THF, AcroSeal™

Product Code. 10198262 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10198262 800 mL AcroSeal™ Glass Bottle 800mL
10759344 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10198262 Supplier Thermo Scientific Acros Supplier No. 209398000
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1826-67-1 | C2H3BrMg | 131.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1826-67-1 , 109-99-9
Molecular Formula C2H3BrMg
Molecular Weight (g/mol) 131.26
MDL Number MFCD00000042
InChI Key XHHHAXOHMKAOSL-UHFFFAOYSA-M
Synonym vinylmagnesium bromide, bromo ethenyl magnesium, vinyl magnesium bromide, magnesium, bromoethenyl, vinylmagnesium bromide solution, vinylmagnesium bromide solution, 1.0 m in thf, bromovinylmagnesium, bromo vinyl magnesium, vinyl magnesiumbromide, grignard reagent
PubChem CID 74584
SMILES Br[Mg]C=C

Specifications

Color Brown
Concentration 0.7 to 0.9M
Density 0.9800g/mL
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.98
Linear Formula H2C=CHMgBr
Quantity 800 mL
Flash Point −17°C
Physical Form Solution
Solubility Information Solubility in water: vigorous reaction.
IUPAC Name magnesium;ethene;bromide
Formula Weight 131.25
Chemical Name or Material Vinylmagnesium bromide, 0.7M Solution in THF
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Vinylmagnesium bromide, 0.7M solution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which, in contact with water, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.