missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VIP Antibody (2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
252.00 € - 684.00 €
Specifications
| Antigen | VIP |
|---|---|
| Clone | 2 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:1000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100 |
| Applications | Immunohistochemistry (Paraffin), Immunohistochemistry, ELISA, Western Blot, Immunofluorescence, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18215074
|
Novus Biologicals
NBP1-05163-0.025ML |
0.025 mL |
252.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18701094
|
Novus Biologicals
NBP1-05163 |
0.1 mL |
684.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VIP Monoclonal specifically detects VIP in Human, Rat, Porcine, Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| VIP | |
| 1.0 mg/mL | |
| Immunohistochemistry (Paraffin), Immunohistochemistry, ELISA, Western Blot, Immunofluorescence, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Cycle and Replication, GPCR, Neuroscience, Neurotransmission, Signal Transduction | |
| P01282 | |
| 7432 | |
| Vasoactive intestinal peptide adsorbed onto aluminum hydroxide gel | |
| Primary | |
| Store at 4C in the dark. |
| 2 | |
| Western Blot 1:100-1:2000, ELISA 1:1000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Bovine, Pig, Rat, Human | |
| MGC13587, PHM27, vasoactive intestinal peptide, VIP peptides | |
| VIP | |
| IgG1 κ | |
| Protein A or G purified | |
| Binds to human, bovine, porcine or rat VIP, but does not bind to PHM or PACAP |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title