missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ VIP Monoclonal Antibody (2)
GREENER_CHOICE

Product Code. 11554132
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
200µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11554132 400 μL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11554132 Supplier Invitrogen™ Supplier No. ABS0230202

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ABS 023-02-02 detects vasoactive intestinal peptide in human, bovine, porcine and rat samples. ABS 023-02-02 has been successfully used in ELISA and Western blot procedures. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.

VIP encodes a protein that belongs to the glucagon family. It stimulates myocardial contractility, causes vasodilation, increases glycogenolysis, lowers arterial blood pressure and relaxes the smooth muscle of trachea, stomach and gall bladder. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VIP
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2
Concentration 1.04 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene VIP
Gene Accession No. P01282, P01283, P01284, P81401
Gene Alias Intestinal peptide PHI-27; Intestinal peptide PHI-42; Intestinal peptide PHM-27; Intestinal peptide PHV-42; MGC13587; PACAP-R-2; Peptide histidine isoleucinamide 27; Peptide histidine methioninamide 27; Peptide histidine valine 42; PHI; PHI, peptide histidine isoleucine; PHM27; prepro-VIP; vasoactive intestinal peptide; Vasoactive intestinal polypeptide; VIP; VIP peptides; vip/phi27; VIP-R-1
Gene Symbols VIP
Host Species Mouse
Immunogen Vasoactive intestinal peptide.
Purification Method Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100500718, 117064, 280956, 7432
Target Species Human, Rat, Bovine, Pig
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.