missing translation for 'onlineSavingsMsg'
Learn More

Vipadenant, MedChemExpress

Product Code. 16490547
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490547 50 mg 50mg
16454208 10 mg 10mg
16426927 5 mg 5mg
16405166 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16490547 Supplier Medchem Express Supplier No. HY10857

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vipadenant (BIIB-014; CEB-4520) is an adenosine receptor antagonist, with Kis of 1.3 nM and 68 nM for A2A and A1, respectively.

Vipadenant (BIIB-014

Specifications

Chemical Name or Material Vipadenant
Physical Form Solid
CAS 442908-10-3
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C16H15N7O
For Use With (Application) Neuroscience-Neuromodulation
Synonym BIIB-014 CEB-4520
Solubility Information DMSO : ≥ 31 mg/mL (96.47 mM)
SMILES NC1=NC2=C(N=NN2CC3=CC=C(N)C(C)=C3)C(C4=CC=CO4)=N1
Molecular Weight (g/mol) 321.34
Formula Weight 321.34
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.