missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Viperin Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93524-0.1ml
This item is not returnable.
View return policy
Description
Viperin Polyclonal antibody specifically detects Viperin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Viperin | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| 2510004L01Rik, cig33, cig5, Cytomegalovirus-induced gene 5 protein, interferon-inducible, radical S-adenosyl methionine domain containing 2, radical S-adenosyl methionine domain-containing protein 2, vig1, Viperin, Virus inhibitory protein, endoplasmic reticulum-associated | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 118-361 of human Viperin (NP_542388.2). EKINFSGGEPFLQDRGEYLGKLVRFCKVELRLPSVSIVSNGSLIRERWFQNYGEYLDILAISCDSFDEEVNVLIGRGQGKKNHVENLQKLRRWCRDYRVAFKINSVINRFNVEEDMTEQIKALNPVRWKVFQCLLIEGENCGEDALREAERFVIGDEEFERFLERHKEVSCLVPESNQKMKDSYLILDEYMRFLNCRKGRKDPSKSILDVGVEEAIKFSGFDEKMFLKRGGKYIWSKADLKLDW | |
| 0.1 mL | |
| Immunology | |
| 91543 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction