missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Viral Dengue Virus 2 NS1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB94392-100
This item is not returnable.
View return policy
Description
Dengue Virus 2 NS1 Monoclonal specifically detects Dengue Virus 2 NS1 in Viral samples. It is validated for ELISA.
Specifications
| Dengue virus 2 NS1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| DEN-2, Dengue NS1, Dengue NS1 protein, Dengue Virus, Dengue virus Serotype 2, Dengue virus Type 2, DENV2, DENV2-NS1 | |
| Human embryonic kidney cell line HEK293-derived recombinant Dengue Virus 2 NS1 protein Asp1-Ala352 Accession # NP_739584 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| ELISA | |
| 969820 | |
| ELISA | |
| NP_739584 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Detects Viral Dengue Virus 2 NS1 in direct ELISAs. | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction