missing translation for 'onlineSavingsMsg'
Learn More

Viral EBV IL-10 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. p-7067773
Change view
Click to view available options
Quantity:
25 μg
500 μg
Unit Size:
25µg
500µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18773898 25 μg 25µg
18169080 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18773898 Supplier R&D Systems Supplier No. MAB9151SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-10 Monoclonal specifically detects IL-10 in Viral samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-10
Applications Western Blot
Classification Monoclonal
Clone 134206
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P03180
Gene Alias CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF
Gene Symbols CeHV15gBCRF1
Host Species Mouse
Immunogen E. coli-derived recombinant viral EBV IL-10 Gln26-Arg170 Accession # P03180
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3586
Test Specificity Detects viral EBV IL-10 in direct ELISAs and Western blots. In Western blots, less than 2% cross-reactivity with IL-10 from canine, cotton rat, feline, human, mouse, porcine, or rat is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Virus
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.