missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Viral MIP-II Monoclonal Antibody (82206)
GREENER_CHOICE

Product Code. 30535636
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30535636 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30535636 Supplier Invitrogen™ Supplier No. MA561310

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Reconstitute at 0.5 mg/mL in sterile PBS.

MIP-II, also known as Macrophage Inflammatory Protein-2 (MIP-2), is a chemokine belonging to the CXC chemokine family. It is primarily produced by macrophages in response to inflammatory stimuli such as bacterial infections, cytokines, and endotoxins. MIP-2 plays a pivotal role in the immune response by attracting and activating neutrophils to sites of infection or injury, thereby contributing to the inflammatory process. This chemokine is involved in various physiological and pathological processes, including wound healing, immune defense, and the development of inflammatory diseases. Elevated levels of MIP-2 are often observed in conditions such as sepsis, arthritis, and chronic obstructive pulmonary disease (COPD), highlighting its significance in disease pathogenesis. Due to its critical role in inflammation and immune response, MIP-2 is a potential target for therapeutic interventions aimed at modulating inflammatory conditions and enhancing host defense mechanisms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Viral MIP-II
Applications Western Blot
Classification Monoclonal
Clone 82206
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with trehalose and no preservative
Host Species Mouse
Immunogen E. coli-derived recombinant human herpes virus-8 MIP-II, Leu24-Arg94.
Purification Method Protein A/G
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.