missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Viral MIP-II Polyclonal Antibody

Description
Reconstitute at 0.2 mg/mL in sterile PBS.
MIP-II, also known as Macrophage Inflammatory Protein-2 (MIP-2), is a chemokine belonging to the CXC chemokine family. It is primarily produced by macrophages in response to inflammatory stimuli such as bacterial infections, cytokines, and endotoxins. MIP-2 plays a pivotal role in the immune response by attracting and activating neutrophils to sites of infection or injury, thereby contributing to the inflammatory process. This chemokine is involved in various physiological and pathological processes, including wound healing, immune defense, and the development of inflammatory diseases. Elevated levels of MIP-2 are often observed in conditions such as sepsis, arthritis, and chronic obstructive pulmonary disease (COPD), highlighting its significance in disease pathogenesis. Due to its critical role in inflammation and immune response, MIP-2 is a potential target for therapeutic interventions aimed at modulating inflammatory conditions and enhancing host defense mechanisms.
Specifications
Specifications
| Antigen | Viral MIP-II |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with trehalose and no preservative |
| Host Species | Goat |
| Immunogen | E. coli-derived recombinant human Viral MIP-II, Leu24-Arg94. |
| Purification Method | Antigen affinity chromatography |
| Quantity | 100 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?