missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Viral Zika Virus (H/PF/2013) Envelope Antibody, R&D Systems™

Mouse Monoclonal Antibody
193.00 € - 431.00 €
Specifications
| Antigen | Zika Virus (H/PF/2013) Envelope |
|---|---|
| Clone | 990801 |
| Dilution | Western Blot 1 μg;/mL, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18752797
![]() |
R&D Systems
MAB10009-SP |
25 μg |
193.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18704117
![]() |
R&D Systems
MAB10009-100 |
100 μg |
431.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Zika Virus (H/PF/2013) Envelope Monoclonal specifically detects Zika Virus (H/PF/2013) Envelope in Viral samples. It is validated for Western Blot, ELISA.Specifications
| Zika Virus (H/PF/2013) Envelope | |
| Western Blot 1 μg;/mL, ELISA | |
| Monoclonal | |
| Lyophilized | |
| Virus | |
| E. coli-derived recombinant Zika Virus (H/PF/2013) Envelope, Leu590-Ser695 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects Zika Virus (H/PF/2013) Envelope (E3) in direct ELISAs. |
| 990801 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title