missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Viral Zika Virus (H/PF/2013) NS1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB99201-100
This item is not returnable.
View return policy
Description
Zika virus (H/PF/2013) NS1 Monoclonal specifically detects Zika virus (H/PF/2013) NS1 in Viral samples. It is validated for ELISA.
Specifications
| Zika virus (H/PF/2013) NS1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2a |
| ELISA | |
| 990907 | |
| ELISA | |
| AHZ13508 | |
| Human embryonic kidney cell, HEK293-derived recombinant Viral Zika Virus (H/PF/2013) NS1, Val796 & Ser799-Ser1148, with a C-terminal 6-His tag, Accession No. AHZ13508 | |
| 100 μg | |
| Detects Viral Zika Virus (H/PF/2013) NS1 in direct ELISAs | |
| Virus | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction