missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Virilizer Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-21363
This item is not returnable.
View return policy
Description
Virilizer Polyclonal specifically detects Virilizer in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).
Specifications
| Virilizer | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| DKFZP434I116, DKFZp781B2117, fSAP121, functional spliceosome-associated protein 121, KIAA1429, MGC138493, MGC141940, protein virilizer homolog | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 25962 | |
| Store at 4C. Do not freeze. |
| Immunoprecipitation, Western Blot | |
| 1.0 mg/mL | |
| Immunoprecipitation 5-10 ug/mg lysate, Western Blot (Negative) | |
| NP_056311.2 | |
| VIRMA | |
| The immunogen recognized by this antibody maps to a region between residue 200 and 250 of human Virilizer (KIAA1429) using the numbering given in entry NP_056311.2 (GeneID 25962). | |
| 100 μL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction