missing translation for 'onlineSavingsMsg'
Learn More

Bolle Safety™ ATOV Visor For Use With Bolle ATOM Safety Goggles

Product Code. 12689090
Change view
Click to view available options
:
ATOV Visor For Use With Bolle ATOM Safety Goggles
Product Code.
12689090 ATOV Visor For Use With Bolle ATOM Safety Goggles
1 options
This item is not returnable. View return policy

Product Code. 12689090

Brand: Bolle Safety™ ATOV

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The ATOV Visor offers additional increased protection for the face when attached to the Bolle ATOM goggles. The polycarbonate visor is compact, lightweight and extremely comfortable thanks to its modular thermo plastic rubber (TPR) frame.

  • Additional protection against chemical liquid droplets
  • Blue polycarbonate
  • Ultra lightweight
  • For use with ATOM safety goggles
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Material Polycarbonate
Certifications/Compliance (Europe) EN 166 3 BT CE
Description Additional visor for use with ATOM safety goggles
Disposable Reusable
Model ATOV
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.