Learn More
VISTA/B7-H5/PD-1H Antibody (CL3981) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-59031PE
Product Code. 30307829
  • 737.00 € / 0.10mL
Description
VISTA/B7-H5/PD-1H Monoclonal antibody specifically detects VISTA/B7-H5/PD-1H in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VISTA/B7-H5/PD-1H
Monoclonal
PE
B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, GI24, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR
This antibody was developed using a recombinant protein derived from Q9H7M9, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Cardiovascular Biology, Immunology
64115
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL3981
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified