missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VISTA Monoclonal Antibody (MIH64), NovaFluor™ Yellow 730, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
178.00 € - 344.00 €
Specifications
| Antigen | VISTA |
|---|---|
| Clone | MIH64 |
| Concentration | 0.1 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30055937
|
Invitrogen™
M035T03Y07-A |
100 μg |
344.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30056013
|
Invitrogen™
M035T02Y07-A |
25 μg |
178.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VISTA Monoclonal antibody specifically detects VISTA in Mouse samples. It is validated for Flow CytometrySpecifications
| VISTA | |
| 0.1 mg/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| Q9D659 | |
| 74048 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| MIH64 | |
| Flow Cytometry | |
| NovaFluor Yellow 730 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| 4632428N05Rik; B7H5; B7-H5; C10orf54; C1H10orf54; C28H10orf54; C4H10orf54; chromosome 10 open reading frame 54; DD1alpha; Death Domain1alpha; Dies1; differentiation of ESCs 1; GI24; PD-1H; PDCD1 homolog; PD-H1; platelet receptor Gi24; PP2135; RIKEN cDNA 4632428N05 gene; SISP1; sisp-1; Stress-induced secreted protein-1; UNQ730/PRO1412; V-domain Ig suppressor of T cell activation; VISTA; V-set domain-containing immunoregulatory receptor; V-set immunoregulatory receptor; VSIR; V-type immunoglobulin domain-containing suppressor of T-cell activation | |
| IgG2a κ | |
| Affinity chromatography | |
| VSIR |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title