missing translation for 'onlineSavingsMsg'
Learn More

Vitamin A acetate 10 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30310786
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30310786 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30310786 Supplier Dr. Ehrenstorfer Supplier No. DREA17923820AL10
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Vitamin A acetate (Retinol acetate)
CAS 127-47-9
Analyte or Component Names Vitamin A acetate (Retinol acetate)
Shipping Condition Room Temperature
Molecular Formula C22 H32 O2
Concentration 10 μg/mL
Certifications/Compliance ISO 17034
Synonym Retinol, acetate, all-trans-, Arovit, Myvak, Myvax, NSC 122045, NSC 122760, Retinol acetate, Retinyl Acetate, Ro 1-5275, Rovimix A 1000, Vitamin A acetate, Vitamin A1 acetate, all-trans-Retinol acetate, all-trans-Retinyl acetate, all-trans-Vitamin A acetate, trans-Retinol acetate, trans-Retinyl acetate
InChI Formula InChI=1S/C22H32O2/c1-17(9-7-10-18(2)14-16-24-20(4)23)12-13-21-19(3)11-8-15-22(21,5)6/h7,9-10,12-14H,8,11,15-16H2,1-6H3/b10-7+,13-12+,17-9+,18-14+
SMILES CC(=O)OC\C=C(/C)\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C
IUPAC Name [(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenyl] acetate
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 328.49
Quantity 1 mL
Formula Weight 328.2402 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.