missing translation for 'onlineSavingsMsg'
Learn More

Vitamin D2, Dr. Ehrenstorfer

Product Code. 30313600
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30313600 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30313600 Supplier Dr. Ehrenstorfer Supplier No. DREC17923900100MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ergocalciferol
CAS 50-14-6
Analyte or Component Names Ergocalciferol
Shipping Condition +20°C
Molecular Formula C28 H44 O
Certifications/Compliance ISO/IEC 17025
Synonym Ergocalciferol, Vitamin D2, 9,10-Secoergosta-5,7,10(19),22-tetraen-3-ol, (3beta,5Z,7E,22E)- (9CI), Cyclohexanol, 4-methylene-3-[2-[tetrahydro-7a-methyl-1-(1,4,5-trimethyl-2-hexenyl)-4(3aH)-indanylidene]ethylidene]- (6CI), Ergocalciferol (7CI,8CI), (1S,3Z)-4-Methylene-3-[(2E)-2-[(1R,3aS,7aR)-octahydro-7a-methyl-1-[(1R,2E,4R)-1,4,5-trimethyl-2-hexen-1-yl]-4H-inden-4-ylidene]ethylidene]cyclohexanol, (+)-Vitamin D2, 9,10-Secoergosta-5,7,10(19),22-tetraen-3beta-ol, Buco-D, Calciferol, Condocaps, Condol, Crystallina, D-Arthin, D-Tracetten, Davitamon D, Davitin, De-Rat Concentrate, Decaps, Dee-Osterol, Dee-Ron, Dee-Ronal, Deltalin, Deratol, Detalup, Diactol, Divit urto, Doral, Drisdol, Ercalciol, Ergorone, Ertron, Fortodyl, Geltabs, Hi-Deratol, Infron, Metadee, Mina D2, Mulsiferol, Mykostin, NSC 62792, Oleovitamin D, Oleovitamin D2, Osteil, Ostelin, Radiostol, Radsterin, Rodine C, Shock-ferol, Sterogyl, Uvesterol D, Vio D, Viosterol, Vitavel-D
InChI Formula InChI=1S/C28H44O/c1-19(2)20(3)9-10-22(5)26-15-16-27-23(8-7-17-28(26,27)6)12-13-24-18-25(29)14-11-21(24)4/h9-10,12-13,19-20,22,25-27,29H,4,7-8,11,14-18H2,1-3,5-6H3/b10-9+,23-12+,24-13-/t20-,22+,25-,26+,27-,28+/m0/s1
SMILES CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2\C(=C\C=C/3\C[C@@H](O)CCC3=C)\CCC[C@]12C
IUPAC Name (1S,3Z)-3-[(2E)-2-[(1R,3aS,7aR)-1-[(E,2R,5R)-5,6-dimethylhept-3-en-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol
Molecular Weight (g/mol) 396.65
Quantity 100 mg
Formula Weight 396.3392 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.