missing translation for 'onlineSavingsMsg'
Learn More

Vitamin D3 10 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30313419
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30313419 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30313419 Supplier Dr. Ehrenstorfer Supplier No. DREA17924000AL10
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Colecalciferol
CAS 67-97-0
Analyte or Component Names Colecalciferol
Shipping Condition Room Temperature
Molecular Formula C27 H44 O
Concentration 10 μg/mL
Certifications/Compliance ISO 17034
Synonym Cyclohexanol, 3-[(2E)-2-[(1R,3aS,7aR)-1-[(1R)-1,5-dimethylhexyl]octahydro-7a-methyl-4H-inden-4-ylidene]ethylidene]-4-methylene-, (1S,3Z)-, 9,10-Secocholesta-5,7,10(19)-trien-3-ol, (3β,5Z,7E)- (9CI), Cholecalciferol (8CI), 9,10-Secocholesta-5,7,10(19)-trien-3β-ol, Aquadetrim, Arachitol, Calciol, Calcirol, Colecalciferol, D3-Vigantol, Decristol, Delsterol, Deparal, Devaron, Devikap, Dry Vitamin D3 100CWS/AM, Duphafral D3 1000, Ekostop D3, FeraCol, Granuvit D3, High-D 2X, Lutavit D, Minisun, NSC 375571, Oleovit D3, Oleovitamin D3, Plivit D3, Quintox, Ricketon, Riken D3 Oil 100, Riken D3 Oil 20, Trivitan, UVEDOSE, Vi-De3, Videkhol, Vigantoletten, Vigorsan, Vitamin D3, Vitamin D3 S 50t, Vitinc Dan-Dee-3
InChI Formula InChI=1S/C27H44O/c1-19(2)8-6-9-21(4)25-15-16-26-22(10-7-17-27(25,26)5)12-13-23-18-24(28)14-11-20(23)3/h12-13,19,21,24-26,28H,3,6-11,14-18H2,1-2,4-5H3/b22-12+,23-13-/t21-,24+,25-,26+,27-/m1/s1
SMILES CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2\C(=C\C=C/3\C[C@@H](O)CCC3=C)\CCC[C@]12C
IUPAC Name (1S,3Z)-3-[(2E)-2-[(1R,3aS,7aR)-7a-methyl-1-[(2R)-6-methylheptan-2-yl]-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 384.64
Quantity 1 mL
Formula Weight 384.3392 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.