missing translation for 'onlineSavingsMsg'
Learn More

Vitamin K{1}

Product Code. 11362866
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11362866 5 g 5g
11352866 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362866 Supplier Thermo Scientific Alfa Aesar Supplier No. L10575.06
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Important dietary source found in green plants, algae and photosynthetic bacteria. It is used as a vitamin (prothrombogenic) and veterinary antidote for dicoumarol poisoning. It is an essential vitamin for the synthesis of coagulation factors II, VII, IX, and X in the liver. | CAS: 84-80-0 | C31H46O2 | 450.707 g/mol

Vitamin K{1} is an important dietary source found in green plants, algae and photosynthetic bacteria. It is used as a vitamin (prothrombogenic) and veterinary antidote for dicoumarol poisoning. It is an essential vitamin for the synthesis of coagulation factors II, VII, IX, and X in the liver.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-80-0
Molecular Formula C31H46O2
Molecular Weight (g/mol) 450.707
MDL Number MFCD00214063
InChI Key MBWXNTAXLNYFJB-NKFFZRIASA-N
Synonym vitamin k1, phytonadione, phylloquinone, phytylmenadione, phyllochinon, phytomenadione, alpha-phylloquinone, 3-phytylmenadione, konakion, 2-methyl-3-phytyl-1,4-naphthoquinone
PubChem CID 5284607
ChEBI CHEBI:18067
IUPAC Name 2-methyl-3-[(E,7R,11R)-3,7,11,15-tetramethylhexadec-2-enyl]naphthalene-1,4-dione
SMILES CC1=C(C(=O)C2=CC=CC=C2C1=O)CC=C(C)CCCC(C)CCCC(C)CCCC(C)C

Specifications

Melting Point ∼-20°C
Density 0.984
Boiling Point ∼140°C (0.001mmHg)
Flash Point >110°C (230°F)
Packaging Glass bottle
Refractive Index 1.526
Quantity 5 g
Beilstein 2568816
Sensitivity Light sensitive
Specific Gravity 0.984
Solubility Information Miscible with dehydrated alcohol,acetone,petroleum ether,hexane,dioxane,chloroform,ether,benzene and vegetable oils. Immiscible with water.
Physical Form Viscous Liquid
Chemical Name or Material Vitamin K1
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.