missing translation for 'onlineSavingsMsg'
Learn More

Vitamin K1, MP Biomedicals™

Product Code. 11417501 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11417501 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11417501 Supplier MP Biomedicals Supplier No. 02103283.2
  • ONLINE EXCLUSIVE PRICE
    43.20 €
    40.95 € / 250mg
    Save 2.25 €
    5% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used as a cofactor. Vitamin K1, MP Biomedicals a fat-soluble vitamin structurally similar to other forms of Vitamin K. It is a cofactor in the formation of coagulation factors II (prothrombin), VII, IX and X and of anticoagulant factors Protein C and S, a

Specifications

CAS 84-80-0
Melting Point -20°C (literature)
Density 0.984 (Lit.)
Boiling Point ∼140°C (literature)
Molecular Formula C31H46O2
Refractive Index 1.523-1.526 (at 25°C)
Quantity 250 mg
Synonym vitamin k1, phytonadione, phylloquinone, phytylmenadione, phyllochinon, phytomenadione, alpha-phylloquinone, 3-phytylmenadione, konakion, 2-methyl-3-phytyl-1,4-naphthoquinone
Solubility Information Soluble in chloroform (clear, yellow solution). Practically insoluble in water, sparingly soluble in alcohol, miscible with fatty oils.
InChI Key MBWXNTAXLNYFJB-NKFFZRIASA-N
SMILES CC1=C(C(=O)C2=CC=CC=C2C1=O)CC=C(C)CCCC(C)CCCC(C)CCCC(C)C
IUPAC Name 2-methyl-3-[(E,7R,11R)-3,7,11,15-tetramethylhexadec-2-enyl]naphthalene-1,4-dione
Molecular Weight (g/mol) 450.707
PubChem CID 5284607
ChEBI CHEBI:18067
Formula Weight 450.7
Physical Form Liquid
Show More Show Less
compliance-icons
Product Identifier
  • Vancomycin Hydrochloride
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P321-Specific treatment (see on this label).
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Vancomycin Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.