missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivaspin™ 20

Product Code. 16646762 Shop All Sartorius Products
Click to view available options
No. per Pack:
12 Pk.
48 Pk.
Unit Size:
Pack of 12
Pack of 48
This item is not returnable. View return policy

Product Code. 16646762

Brand: Sartorius VS2001IVD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Vivaspin™ 20 combines twin vertical membranes and thin flow channel design for high process speeds and concentration factors. It also offers unique pressure and pressure-fuge operation for sensitive samples and even faster processing.

  • CE certified according to EU 98/79/EC IVDD
  • Easy-to-use centrifugal or solvent absorption devices
  • No risk of sample cross-contamination
  • Fast concentration with minimal membrane fouling
  • High sample recoveries for increased sensitivity
  • Printed graduations on the concentrator body provide a convenient indication of retentate volume and simple, direct recovery from the integral dead-stop pocket is possible with standard pipettes.
  • Typical Applications:
    • Concentration of patient samples prior to electrophoresis or immuno-electrophoresis
    • Concentration of urine, cerebrospinal fluid, or plasma to detect disease markers such as Bence Jones Protein
    • Deproteinization of blood samples to aid detection of disease metabolites
TRUSTED_SUSTAINABILITY

Specifications

Material Polycarbonate
Material (Membrane) PES
MWCO 10 kDa
Sterility Non-sterile
For Use With (Application) For in vitro diagnostics (IVD) applications.
No. per Pack 12 Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.