missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VNN2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93907-0.1ml
This item is not returnable.
View return policy
Description
VNN2 Polyclonal antibody specifically detects VNN2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| VNN2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| EC 3.5.1, EC 3.5.1.92, FOAP-4, Glycosylphosphatidyl inositol-anchored protein GPI-80, GPI-80, GPI-80 variant protein 1, GPI-80 variant protein 2, GPI-80 variant protein 3, GPI-80 variant protein 4, pantetheinase, Protein FOAP-4, vanin 2, vanin-2, Vannin 2, vascular non-inflammatory molecule 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 270-493 of human VNN2 (NP_004656.2). VSLNMTGSGIYAPNGPKVYHYDMKTELGKLLLSEVDSHPLSSLAYPTAVNWNAYATTIKPFPVQKNTFRGFISRDGFNFTELFENAGNLTVCQKELCCHLSYRMLQKEENEVYVLGAFTGLHGRRRREYWQVCTMLKCKTTNLTTCGRPVETASTRFEMFSLSGTFGTEYVFPEVLLTEIHLSPGKFEVLKDGRLVNKNGSSGPILTVSLFGRWYTKDSLYSSC | |
| 0.1 mL | |
| Endocrinology, Signal Transduction, Stem Cells | |
| 8875 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction