missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VOC-Mix 61 1000-10000 μg/mL in Methanol, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| CAS | 71-55-6, 107-06-2, 75-27-4, 67-66-3, 156-59-2, 124-48-1 |
| Quantity | 1 mL |
| Concentration | Tetrachloroethene 999.8 μg/mL∣Chloroform 1000 μg/mL∣Tribromomethane 1000 μg/mL∣Trichloroethene 1000 μg/mL∣Bromodichloromethane 1000.3 μg/mL∣Dibromochloromethane 1000.3 μg/mL∣Tetrachloromethane 1000.5 μg/mL∣1,1,1-Trichloroethane 1001.1 μg/mL∣Dichloromethane (Methylenechloride) 5007.2 μg/mL∣1,2-Dichloroethane 10002.3 μg/mL∣cis-1,2-Dichloroethene 10005.1 μg/mL |
| Certifications/Compliance | ISO/IEC 17025 |
| Solvent or Matrix | Methanol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction