missing translation for 'onlineSavingsMsg'
Learn More

Volinanserin, MedChemExpress

Product Code. p-200097560
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16482387

Brand: Medchem Express HY14940

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Volinanserin is a potent and selective antagonist of 5-HT2 receptor, with a Ki of 0.36 nM, and shows 300-fold selectivity for 5-HT2 receptor over 5-HT1c, alpha-1 and DA D2 receptors. Volinanserin has antipsychotic activity.

Volinanserin is a potent and selective antagonist of 5-HT2 receptor, with a Ki of 0.36 nM, and shows 300-fold selectivity for 5-HT2 receptor over 5-HT1c, alpha-1 and DA D2 receptors. Volinanserin has antipsychotic activity.

Specifications

Chemical Name or Material Volinanserin
CAS 139290-65-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H28FNO3
Quantity 50 mg
Synonym MDL100907 M 100907
Solubility Information DMSO : 50 mg/mL (133.88 mM; Need ultrasonic)
SMILES FC1=CC=C(CCN2CCC([C@@]([H])(O)C3=CC=CC(OC)=C3OC)CC2)C=C1
Molecular Weight (g/mol) 373.46
Formula Weight 373.46
Percent Purity 98.12%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.