missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Von Willebrand Factor Antibody (VWF/7979R), Novus Biologicals™
Rabbit Monoclonal Antibody
298.00 € - 530.00 €
Specifications
| Antigen | Von Willebrand Factor |
|---|---|
| Clone | VWF/7979R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18662124
|
Novus Biologicals
NBP3-20322-100ug |
100 μg |
530.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18602694
|
Novus Biologicals
NBP3-20322-20ug |
20 μg |
298.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Von Willebrand Factor Monoclonal antibody specifically detects Von Willebrand Factor in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Von Willebrand Factor | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| 10mM PBS with 0.05% BSA | |
| 7450 | |
| IgG κ | |
| Protein A or G purified |
| VWF/7979R | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| coagulation factor VIII VWF, F8, F8VWF, von Willebrand factor, VWD, vWF | |
| Recombinant fragment (around aa 1150-1250) of human Von Willebrand Factor protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title