missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Von Willebrand Factor Rabbit anti-Human, Clone: 111, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89724-100UL
This item is not returnable.
View return policy
Description
Von Willebrand Factor Monoclonal specifically detects Von Willebrand Factor in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA Detection.
Specifications
| Von Willebrand Factor | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 111 | |
| ELISA 1:5000-1:10000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500-1:2000, Sandwich ELISA Detection 1:1000-1:10000 | |
| coagulation factor VIII VWF, F8, F8VWF, von Willebrand factor, VWD, vWF | |
| Recombinant Human Von Willebrand Factor protein | |
| 100 μg | |
| Cancer | |
| 7450 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction