Learn More
Fisherbrand™ Vortex Mixers Tube Holders
Product Code. 15776959 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
No. of Positions:
1
18
24
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Positions Unit Size
15776959 1 Each
15746959 24 Each
15756959 18 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15776959
  • ONLINE EXCLUSIVE PRICE
    120.00 €
    99.93 € / Pack of 1
    Save 20.07 €
    16% Off
Estimated Shipment: 23-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use with Fisherbrand vortex mixers
  • Mount atop mixer for single-tube, hands-free mixing
  • Easily attach without tools
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Vortex mixer, single tube
No. of Positions 1
Packaging Quantity 1 Ea.