missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS11 Antibody (SR1048), Novus Biologicals™
Rabbit Monoclonal Antibody
338.00 € - 449.00 €
Specifications
| Antigen | VPS11 |
|---|---|
| Clone | SR1048 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:80, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18640194
|
Novus Biologicals
NBP3-22131-100ul |
100 μg |
449.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18669913
|
Novus Biologicals
NBP3-22131-50ul |
50 μg |
338.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VPS11 Monoclonal antibody specifically detects VPS11 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ImmunohistochemistrySpecifikationer
| VPS11 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:80, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| hVPS11, PEP5, RING finger protein 108, RNF108END1, vacuolar protein sorting 11 (yeast homolog), vacuolar protein sorting 11 homolog (S. cerevisiae), vacuolar protein sorting protein 11, vacuolar protein sorting-associated protein 11 homolog | |
| A synthesized peptide derived from human VPS11 (Uniprot #: Q9H270) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1048 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Autophagy, Zinc Finger | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 55823 | |
| IgG | |
| Affinity purified |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel