missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS13B Monoclonal Antibody (OTI6F6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807801S
This item is not returnable.
View return policy
Description
VPS13B Monoclonal antibody specifically detects VPS13B in Human samples. It is validated for Western BlotSpecifications
| VPS13B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q14956 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10457 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI6F6 | |
| Unconjugated | |
| GPNMB | |
| 1810042B05Rik; 2310042E16Rik; 4732488H20; C330002D13Rik; C87206; CHS1; COH1; Cohen syndrome homolog 1; Cohen syndrome protein 1; Cohen syndrome protein 1 homolog; D230005K13; KIAA0532; mKIAA0532; mKIAA5032; vacuolar protein sorting 13 homolog B; vacuolar protein sorting 13B (yeast); vacuolar protein sorting-associated protein 13B; Vps13b | |
| Full length human recombinant protein of human GPNMB produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction