missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VPS18 Recombinant Rabbit Monoclonal Antibody (24GB2405)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557320
This item is not returnable.
View return policy
Description
VPS18 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
VPS18 is a vesicle mediated protein that plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene encodes the human homolog of yeast class C Vps18 protein. The mammalian class C Vps proteins are predominantly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway.
Specifications
| VPS18 | |
| Recombinant Monoclonal | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8R307, Q9P253 | |
| VPS18 | |
| A synthesized peptide derived from human VPS18 (aa 1-28). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB2405 | |
| Unconjugated | |
| VPS18 | |
| 9930024E13Rik; hVPS18; KIAA1475; PEP3; vacuolar protein sorting 18 (yeast); vacuolar protein sorting 18 homolog; vacuolar protein sorting 18 homolog (S. cerevisiae); vacuolar protein sorting protein 18; vacuolar protein sorting-associated protein 18 homolog; Vps18; VPS18 CORVET/HOPS core subunit; VPS18, CORVET/HOPS core subunit | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 228545, 296083, 57617 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction