Learn More
Invitrogen™ VPS18 Recombinant Rabbit Monoclonal Antibody (24GB2405)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557319
 View more versions of this product
Product Code. 30276024
  • 500.00 € / 100µL
Description
VPS18 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
VPS18 is a vesicle mediated protein that plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene encodes the human homolog of yeast class C Vps18 protein. The mammalian class C Vps proteins are predominantly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VPS18
Recombinant Monoclonal
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q8R307, Q9P253
VPS18
A synthesized peptide derived from human VPS18 (aa 1-28).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB2405
Unconjugated
VPS18
9930024E13Rik; hVPS18; KIAA1475; PEP3; vacuolar protein sorting 18 (yeast); vacuolar protein sorting 18 homolog; vacuolar protein sorting 18 homolog (S. cerevisiae); vacuolar protein sorting protein 18; vacuolar protein sorting-associated protein 18 homolog; Vps18; VPS18 CORVET/HOPS core subunit; VPS18, CORVET/HOPS core subunit
Rabbit
Affinity chromatography
RUO
228545, 296083, 57617
-20°C