missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD266 Recombinant Rabbit Monoclonal Antibody (24GB2070)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557789
This item is not returnable.
View return policy
Description
CD266 Recombinant Monoclonal Antibody for Western Blot, Flow
TWEAK is a member of the TNF family and was first described as a weak inducer of apoptosis in demonstrate that this relatively small molecule is a fully transformed cell lines. Recently, a receptor for TWEAK was isolated by expression cloning from a HUVEC cell cDNA library. TweakR is a small protein belonging to TNF receptor super family. TweakR is capable of intiating a proliferative signal by binding to TRAF1, 2, and 3. TweakR mRNA is upregulated during tissue regeneration in the aorta and liver. It may also play a role in vascular cell migration and angiogenesis.
Specifications
| CD266 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| TNFRSF12A | |
| AI255180; C87282; CD266; FGF-inducible 14; Fgfrp2; fibroblast growth factor regulated protein 2; fibroblast growth factor-inducible immediate-early response protein 14; Fibroblast growth factor-regulated protein 2; FN14; HPIP; TNF receptor superfamily member 12A; TNFRSF12A; Tumor necrosis factor receptor superfamily member 12A; tumor necrosis factor receptor superfamily, member 12A; TweakR; TWEAK-R; Tweak-receptor; type I transmembrane protein Fn14 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 51330 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB2070 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9NP84 | |
| TNFRSF12A | |
| A synthesized peptide derived from human TWEAKR (aa 81-129). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction