missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS26A Antibody (CL2287), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
353.00 € - 518.00 €
Specifications
| Antigen | VPS26 |
|---|---|
| Clone | CL2287 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18727903
|
Novus Biologicals
NBP2-36754 |
0.1 mL |
518.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18655585
|
Novus Biologicals
NBP2-36754-25ul |
25 μL |
353.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VPS26A Monoclonal specifically detects VPS26A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| VPS26 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Autophagy | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| FLJ12930, HB58, Hbeta58, hVPS26, PEP8A, vacuolar protein sorting 26 (yeast homolog), vacuolar protein sorting 26 homolog A (S. pombe), vacuolar protein sorting 26 homolog A (yeast), vacuolar protein sorting-associated protein 26A, Vesicle protein sorting 26A, VPS26vacuolar protein sorting 26 (yeast) | |
| VPS26A | |
| IgG1 | |
| Protein A purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| CL2287 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| O75436 | |
| 9559 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LRKQRTNFHQRFESPESQASAEQPEM | |
| Primary | |
| Protein A purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title