missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VPS26A Recombinant Rabbit Monoclonal Antibody (24GB2415)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557843
This item is not returnable.
View return policy
Description
VPS26A Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene belongs to a group of vacuolar protein sorting (VPS) genes. The encoded protein is a component of a large multimeric complex, termed the retromer complex, involved in retrograde transport of proteins from endosomes to the trans-Golgi network. The close structural similarity between the yeast and human proteins that make up this complex suggests a similarity in function. Expression studies in yeast and mammalian cells indicate that this protein interacts directly with VPS35, which serves as the core of the retromer complex. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
| VPS26A | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Vps26a | |
| AA407240; FLJ12930; H beta 58; H>beta>58 protein; HB58; Hbeta58; Hbeta58 homolog; hVPS26; mVPS26; OTTHUMP00000019721; PEP8A; vacuolar protein sorting 26 A; vacuolar protein sorting 26 homolog A; vacuolar protein sorting 26 homolog A (S. pombe); vacuolar protein sorting 26 homolog A (yeast); vacuolar protein sorting-associated protein 26A; vacuole protein sorting 26; vesicle protein sorting 26A; Vps26; VPS26 retromer complex comonent A; VPS26 retromer complex component A; VPS26, retromer complex component A; VPS26A | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 30930, 361846, 9559 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB2415 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75436, P40336, Q6AY86 | |
| Vps26a | |
| A synthesized peptide derived from human VPS26 (aa 270-327). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction