missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VPS28 Recombinant Rabbit Monoclonal Antibody (24GB1755)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557845
This item is not returnable.
View return policy
Description
VPS28 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
This gene encodes a protein involved in endosomal sorting of cell surface receptors via a multivesicular body/late endosome pathway. The encoded protein is one of the three subunits of the ESCRT-I complex (endosomal complexes required for transport) involved in the sorting of ubiquitinated proteins. The two other subunits of ESCRT-I are vesicular protein sorting 23, also known as tumor susceptibility gene 101 (TSG101), and vesicular protein sorting 37. Two alternative transcripts encoding different isoforms have been described. Additional alternative transcripts may exist but the proteins encoded by these transcripts have not been verified experimentally.
Specifications
| VPS28 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| VPS28 | |
| 1110014J03Rik; Caspase-activated DNase inhibitor that interacts with ASK1; CIIA; D730005C08Rik; ESCRT-I complex subunit VPS28; H-Vps28; MGC60323; vacuolar protein sorting 28 (yeast); vacuolar protein sorting 28 homolog; vacuolar protein sorting 28 homolog (S. cerevisiae); vacuolar protein sorting 28-like protein; vacuolar protein sorting-associated protein 28 homolog; VPS28; VPS28 subunit of ESCRT-I; VPS28, ESCRT-I subunit; yeast class E protein Vps28p homolog | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 300052, 51160, 66914 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1755 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| B5DEN9, Q9D1C8, Q9UK41 | |
| VPS28 | |
| A synthesized peptide derived from human beta VPS28 (aa 150-221). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction