Learn More
Invitrogen™ ROBO1 Recombinant Rabbit Monoclonal Antibody (HL2559)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA556864
Product Code. 30275740
  • 500.00 € / 100µL
Description
ROBO1 Recombinant Monoclonal Antibody for Western Blot, IHC (P)
Specialized cells at the midline, which separates the left and right halves of the CNS, have a number of roles in directing growth cone behavior. In the vertebrate spinal cord, the insect ventral nerve cord and in C. elegans, midline cells produce guidance cues such as nectins and slit, which act as attractants and repellents, respectively. These cells may act as gatekeepers to prevent axons from crossing the midline and to induce a switch in growth cone responsiveness to guidance cues beyond the gateway. One such gatekeeper, Robo, is an axon guidance receptor that defines a novel subfamily of Ig superfamily proteins that are conserved from fruit flies to mammals. Robo acts as a receptor for the repellent Slit and functions in a cell-autonomous fashion. Non-crossing axons express high levels of Robo, whereas crossing axons express low levels of Robo before reaching the midline and high levels after they cross. Robo1 and Robo2 are two human homologs of the Drosophila protein Roundabout. Robo1 is also homologous to the C. elegans gene sax3, whereas Robo2 is homologous to the zebrafish gene astray.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ROBO1
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
O55005, O89026, Q9Y6N7
ROBO1
Recombinant protein encompassing a sequence within the Extracellular domain of human ROBO1. The exact sequence is proprietary.
100 μL
Primary
Human, Mouse, Rat, Zebrafish
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot
HL2559
Unconjugated
ROBO1
AW494633; AW742721; deleted in U twenty twenty; DUTT1; FLJ21882; Gm310; HGNC:10249; H-Robo-1; MGC131599; MGC133277; ROBO1; roundabout 1; roundabout guidance receptor 1; roundabout homolog 1; roundabout homolog 1 (Drosophila); roundabout, axon guidance receptor, homolog 1; roundabout, axon guidance receptor, homolog 1 (Drosophila); Roundabout1 protein; SAX3
Rabbit
Protein A
RUO
19876, 30769, 58946, 6091
-20°C, Avoid Freeze/Thaw Cycles