Learn More
VPS34 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
Brand:  Novus Biologicals NB110-87320
 View more versions of this product
Product Code. 18789563
  • 531.00 € / 0.10mL
Description
VPS34 Polyclonal specifically detects VPS34 in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence, Knockdown Validated.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyclonal
100 kDa
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot 1:1000, Simple Western 1:20, Immunocytochemistry/Immunofluorescence 1:100, Knockdown Validated
Pig