missing translation for 'onlineSavingsMsg'
Learn More

VR23, MedChemExpress

Product Code. 16493627
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493627 50 mg 50mg
16487526 100 mg 100mg
16427618 10 mg 10mg
16420088 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16493627 Supplier Medchem Express Supplier No. HY18741

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress VR23 is a small molecule that potently inhibits the activities of trypsin-like proteasomes (IC50=1 nM), chymotrypsin-like proteasomes (IC50=50-100 nM), and caspase-like proteasomes (IC50=3 μM).

VR23 is a small molecule that potently inhibits the activities of trypsin-like proteasomes (IC50=1 nM), chymotrypsin-like proteasomes (IC50=50-100 nM), and caspase-like proteasomes (IC50=3 μM).

Specifications

Chemical Name or Material VR23
CAS 1624602-30-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C19H16ClN5O6S
Quantity 50 mg
Solubility Information DMSO : 33.33 mg/mL (69.75 mM; Need ultrasonic)
SMILES O=S(N1CCN(C2=CC=NC3=CC(Cl)=CC=C23)CC1)(C4=CC=C([N+]([O-])=O)C=C4[N+]([O-])=O)=O
Molecular Weight (g/mol) 477.88
Formula Weight 477.88
Percent Purity 98.05%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.