missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VSIG8 Monoclonal Antibody (961829)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA562026
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
VSIG8 contains 2 Ig-like V-type (immunoglobulin-like) domains. VSIG8 is single-pass type I membrane protein. The function of the VSIG8 protein remains unknown.
Specifications
| VSIG8 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P0DPA2 | |
| VSIG8 | |
| Human embryonic kidney cell line HEK293-derived recombinant human VSIG8, Met1-Gly263. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Inhibition Assays, Neutralization | |
| 961829 | |
| Unconjugated | |
| VSIG8 | |
| A030011M19; C1orf204; EG240916; LOC100727929; LOW QUALITY PROTEIN: V-set and immunoglobulin domain-containing protein 8; RGD1562464; V-set and immunoglobulin domain containing 8; V-set and immunoglobulin domain containing 8; V-set and immunoglobulin domain-containing protein 8; V-set and immunoglobulin domain-containing protein 8; Vsig8 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 391123 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction