missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VTI1B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00 € - 326.00 €
Specifications
| Antigen | VTI1B |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1-4 μL/mg of lysate |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18633272
|
Novus Biologicals
NBP2-96993-100ul |
100 μg |
326.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18673602
|
Novus Biologicals
NBP2-96993-50ul |
50 μg |
216.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VTI1B Polyclonal antibody specifically detects VTI1B in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| VTI1B | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 10490 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-4 μL/mg of lysate | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| vesicle transport through interaction with t-SNAREs homolog 1B (yeast), Vesicle transport v-SNARE protein Vti1-like 1, vesicle-associated soluble NSF attachment protein receptor, v-SNARE, VTI1, VTI1L, VTI1L1, VTI1-LIKE, Vti1-rp1, VTI2vesicle transport through interaction with t-SNAREs homolog 1B | |
| Produced in rabbits immunized with purified, recombinant Human VTI1B (rh VTI1B; NP_006361.1; Ala 2-Leu 208) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title