missing translation for 'onlineSavingsMsg'
Learn More

VTP-27999 TFA, MedChemExpress

Product Code. p-200101763
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16448108

Brand: Medchem Express HY50769

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress VTP-27999 TFA is an alkyl amine Renin inhibitor; VTP-27999 TFA is useful for Hypertension and End-Organ Diseases.

VTP-27999 TFA is an alkyl amine Renin inhibitor

Specifications

Chemical Name or Material VTP-27999 TFA
Physical Form Oil
CAS 1013937-63-7
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C28H42ClF3N4O7
For Use With (Application) COVID-19-immunoregulation
Solubility Information H2O : 10 mg/mL (15.65 mM; Need ultrasonic) ∣Ethanol : 2.4 mg/mL (3.76 mM; Need ultrasonic)
SMILES O=C(O)C(F)(F)F.COC(NCCO[C@@H](C1=CC(Cl)=CC=C1)[C@@]2([H])CN(CCC2)C(NC[C@@H](NC)C[C@H]3CCCOC3)=O)=O
Molecular Weight (g/mol) 639.1
Formula Weight 639.1
Percent Purity 97.2%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.