missing translation for 'onlineSavingsMsg'
Learn More

VU 0240551, MedChemExpress

Product Code. 16497128
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497128 10 mg 10mg
16479667 5 mg 5mg
16403167 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16497128 Supplier Medchem Express Supplier No. HY16689
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress VU 0240551 is a potent neuronal K-Cl cotransporter KCC2 inhibitor (IC50=560 nM) and is selective versus NKCC1. VU 0240551 also inhibits hERG and L-type Ca2+ channels. VU 0240551 attenuates GABA-induced hyperpolarization of P cells, produces a positive shift in the P cell GABA reversal potential and enhances P cell synaptic transmission.

VU 0240551 is a potent neuronal K-Cl cotransporter KCC2 inhibitor (IC50=560 nM) and is selective versus NKCC1. VU 0240551 also inhibits hERG and L-type Ca2+ channels. VU 0240551 attenuates GABA-induced hyperpolarization of P cells, produces a positive shift in the P cell GABA reversal potential and enhances P cell synaptic transmission.

Specifications

Chemical Name or Material VU 0240551
Physical Form Solid
CAS 893990-34-6
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C16H14N4OS2
Solubility Information DMSO : ≥ 50 mg/mL (146.01 mM)
SMILES O=C(CSC1=NN=C(C2=CC=CC=C2)C=C1)NC3=NC(C)=CS3
Molecular Weight (g/mol) 342.44
Formula Weight 342.44
Percent Purity 99.52%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.