missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VWA8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
288.00 € - 572.00 €
Specifications
| Antigen | KIAA0564 |
|---|---|
| Dilution | Western Blot 1:100 - 1:250, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18420972
|
Novus Biologicals
NBP2-30596-25ul |
25 μL |
288.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18141863
|
Novus Biologicals
NBP2-30596 |
0.1 mL |
572.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VWA8 Polyclonal specifically detects VWA8 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| KIAA0564 | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| FLJ21779, hypothetical protein LOC23078, KIAA0564 | |
| VWA8 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 1:100 - 1:250, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Human | |
| A3KMH1 | |
| 23078 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: EHAIKEIVKEEADEYFVIVLSDANLSRYGIHPAKFAQILTRDPQVNAFAIFIGSLGDQATRLQRTLPAGRSFVAMDTKDIPQILQQIFTSTML | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title