missing translation for 'onlineSavingsMsg'
Learn More
Learn More
vWF-A2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
231.00 € - 326.00 €
Specifications
| Antigen | vWF-A2 |
|---|---|
| Dilution | ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18624838
|
Novus Biologicals
NBP3-06544-100ul |
100 μg |
326.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18627417
|
Novus Biologicals
NBP3-06544-50ul |
50 μg |
231.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
vWF-A2 Polyclonal antibody specifically detects vWF-A2 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
| vWF-A2 | |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| F8VWF, VWD, VWF von Willebrand factor | |
| Produced in rabbits immunized with purified, recombinant Human vWF-A2 (Accession#: AAB59458.1; Met1-Lys2813) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, pH7.0 | |
| 7450 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title