missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Vybrant™ DyeCycle™ Violet Ready Flow™ Reagent

Product Code. 15942344
Change view
Click to view available options
Quantity:
120 reactions
Unit Size:
120 reactions
Product Code. Quantity unitSize
15942344 120 reactions 120 reactions
1 options
This item is not returnable. View return policy

Product Code. 15942344

Brand: Invitrogen™ R37172

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Vybrant DyeCycle Violet Ready Flow Reagent is a bright, easy-to-use, double-stranded DNA stain for cell cycle analysis in living cells using flow cytometry.

Vybrant DyeCycle Violet Ready Flow Reagent is a bright, easy-to-use, double-stranded DNA stain for cell cycle analysis in living cells using flow cytometry. It is excitable by a 405-nm laser with an emission peak at 437 nm.

• Stable at room temperature
• Convenient, ready-to-use format—no need to dilute, weigh, or pipette
• Accurate cell-cycle analysis in living cells
• Low toxicity for cell sorting and additional live cell experiments

Vybrant DyeCycle Violet Ready Flow Reagent is a ready-to-use, direct-to-sample DNA stain. Add 2 drops from the dropper bottle directly into 1 x 106 cells, incubate, and analyze.

As a cell-permeable dye, Vybrant DyeCycle Violet Ready Flow enters live cells and selectively binds dsDNA and emits a signal proportional to the DNA mass. The dye can be detected when excited with a 405-nm laser, and the emission captured at a peak of 437 nm. Vybrant DyeCycle Violet reagent is an ideal choice for DNA content analysis and can be included in multi-color cell cycle studies.

TRUSTED_SUSTAINABILITY

Specifications

Color Violet
Content And Storage 2 dropper bottles, store at room temperature
Product Type Ready Flow Reagent
Emission 369/437
Product Line Ready Flow, Vybrant
Quantity 120 reactions
Shipping Condition Room Temperature
Sub Cellular Localization Nucleus

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.